siRNA Details
| Gene Symbol: Lrriq4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290510.1 | NP_001277439.1 | 2 | 1074 | ||
| NM_026668.2 | NP_080944.2 | 4 | 1237 | |||
| GenBank mRNA | AK015024.1 | - | 2 | 1025 | ||
| AK019775.1 | - | 4 | 1238 | |||
| BC145806.1 | - | 4 | 938 | 40131109 | ||

Lrriq4
Genomic Map