siRNA Details
| Gene Symbol: ARID3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001307939.1 | NP_001294868.1 | 2 | 528 | ||
| NM_006465.3 | NP_006456.1 | 2 | 528 | |||
| GenBank mRNA | AB208830.1 | - | 2 | 447 | ||
| AF116846.1 | - | 2 | 460 | |||
| AK298716.1 | - | 2 | 453 | |||
| BC041792.1 | - | 2 | 456 | 5297774 | ||
| BC045545.1 | - | 2 | 502 | |||
| BC060824.1 | - | 2 | 441 | |||

ARID3B
Genomic Map