siRNA Details
| Gene Symbol: Ccdc88c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026681.4 | NP_080957.2 | 23, 24 | 4237 | ||
| GenBank mRNA | AK173188.1 | - | 9, 10 | 1612 | ||
| AY339881.1 | - | 23, 24 | 4238 | |||
| BC043115.1 | - | 25, 26 | 3848 | 6408630 | ||
| BC063255.1 | - | 25, 26 | 3848 | 6408630 | ||

Ccdc88c
Genomic Map