siRNA Details
| Gene Symbol: Ccdc163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026714.2 | NP_080990.2 | 6 | 662 | ||
| GenBank mRNA | AI606993.1 | - | 2 | 194 | ||
| AK002768.1 | - | 6 | 564 | |||
| BC022718.1 | - | 5 | 661 | |||
| BC022733.1 | - | 6 | 685 | 4506628 | ||

Ccdc163
Genomic Map