siRNA Details
| Gene Symbol: Ccdc163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026714.2 | NP_080990.2 | 10 | 1070 | ||
| GenBank mRNA | AK002768.1 | - | 10 | 972 | ||
| BC022718.1 | - | 9 | 1069 | |||
| BC022733.1 | - | 10 | 1093 | 4506628 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc163| Gene Symbol: Ccdc163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026714.2 | NP_080990.2 | 10 | 1070 | ||
| GenBank mRNA | AK002768.1 | - | 10 | 972 | ||
| BC022718.1 | - | 9 | 1069 | |||
| BC022733.1 | - | 10 | 1093 | 4506628 | ||