siRNA Details
| Gene Symbol: Lmbrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026719.2 | NP_080995.2 | 7 | 805 | ||
| GenBank mRNA | AK032560.1 | - | 1104 | |||
| AK152170.1 | - | 7 | 616 | |||
| AK153237.1 | - | 7 | 616 | |||
| AK158982.1 | - | 7 | 641 | |||
| AK167080.1 | - | 7 | 624 | |||
| AY061989.1 | - | 6 | 516 | |||
| BC025579.1 | - | 7 | 637 | 5325113 | ||
| BC027774.1 | - | 7 | 688 | 5346846 | ||
| BC031902.1 | - | 7 | 628 | 5362166 | ||
| BC039053.1 | - | 7 | 719 | 2655321 | ||

Lmbrd1
Genomic Map