siRNA Details
| Gene Symbol: Ints1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026748.2 | NP_081024.3 | 17, 18 | 2618 | ||
| GenBank mRNA | AB257854.1 | - | 17, 18 | 2578 | ||
| BC063266.1 | - | 17, 18 | 2605 | 6409930 | ||
| CB526682.1 | - | 4, 5 | 337 | |||

Ints1
Genomic Map