siRNA Details
| Gene Symbol: 2310036O22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026760.2 | NP_081036.2 | 3 | 725 | ||
| GenBank mRNA | AK003299.1 | - | 3 | 690 | ||
| AK009653.1 | - | 3 | 692 | |||
| BC023284.1 | - | 3 | 735 | 3484935 | ||
| BC029622.1 | - | 3 | 725 | 4207296 | ||

2310036O22Rik
Genomic Map