siRNA Details
| Gene Symbol: Pmvk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026784.3 | NP_081060.2 | 5 | 1165 | ||
| NM_027348.1 | NP_081624.1 | 5 | 1134 | |||
| GenBank mRNA | AK003607.1 | - | 5 | 968 | ||
| AK013477.1 | - | 5 | 1132 | |||
| AK150483.1 | - | 5 | 1084 | |||
| AK168358.1 | - | 5 | 1060 | |||
| BC028659.1 | - | 4 | 953 | 1247653 | ||

Pmvk
Genomic Map