siRNA Details
| Gene Symbol: Fbxw9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026791.2 | NP_081067.2 | 6 | 1000 | ||
| GenBank mRNA | AK003747.1 | - | 1 | 64 | ||
| AK082085.1 | - | 5 | 1489 | |||
| AK156709.1 | - | 5 | 1150 | |||
| BC043658.1 | - | 6 | 961 | |||
| BC053694.1 | - | 4 | 416 | |||
| BC060082.1 | - | 3 | 231 | |||
| BC094896.1 | - | 1 | 443 | |||
| BC144773.1 | - | 6 | 1366 | |||
| BC145661.1 | - | 6 | 1366 | |||
| CF197934.1 | - | 4 | 404 | |||

Fbxw9
Genomic Map