siRNA Details
| Gene Symbol: Gtf2f2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026816.3 | NP_081092.1 | 3, 4 | 287 | ||
| GenBank mRNA | AK144858.1 | - | 3, 4 | 288 | ||
| BC027173.1 | - | 3, 4 | 283 | 3986466 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gtf2f2| Gene Symbol: Gtf2f2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026816.3 | NP_081092.1 | 3, 4 | 287 | ||
| GenBank mRNA | AK144858.1 | - | 3, 4 | 288 | ||
| BC027173.1 | - | 3, 4 | 283 | 3986466 | ||