siRNA Details
| Gene Symbol: Disp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278218.1 | NP_001265147.1 | 9 | 3055 | ||
| NM_001278219.1 | NP_001265148.1 | 9 | 3015 | |||
| NM_001278220.1 | NP_001265149.1 | 8 | 2898 | |||
| NM_026866.3 | NP_081142.3 | 10 | 3143 | |||
| GenBank mRNA | AK135755.1 | - | 1 | 950 | ||
| AK138276.1 | - | 10 | 3100 | |||
| AK170113.1 | - | 9 | 3010 | |||
| AY144589.1 | - | 8 | 2853 | |||
| AY150577.1 | - | 8 | 2802 | |||
| AY150698.1 | - | 10 | 3035 | |||
| BC043102.1 | - | 9 | 3016 | 6406376 | ||
| BC059225.1 | - | 9 | 3016 | 6406376 | ||

Disp1
Genomic Map