siRNA Details
| Gene Symbol: Ubap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026872.1 | NP_081148.1 | 5, 6 | 571 | ||
| GenBank mRNA | AF276965.1 | - | 4, 5 | 475 | ||
| BC007179.1 | - | 5, 6 | 571 | 3496608 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ubap2| Gene Symbol: Ubap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026872.1 | NP_081148.1 | 5, 6 | 571 | ||
| GenBank mRNA | AF276965.1 | - | 4, 5 | 475 | ||
| BC007179.1 | - | 5, 6 | 571 | 3496608 | ||