siRNA Details
| Gene Symbol: Chmp2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026879.2 | NP_081155.1 | 6 | 1547 | ||
| GenBank mRNA | AK004506.1 | - | 3 | 1020 | ||
| AK084111.1 | - | 6 | 1549 | |||
| AK206871.1 | - | 1 | 330 | |||
| BC042626.1 | - | 5 | 1303 | |||
| BC055809.1 | - | 6 | 1576 | 6414077 | ||

Chmp2b
Genomic Map