siRNA Details
| Gene Symbol: Dcaf12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026893.3 | NP_081169.2 | 2 | 529 | ||
| GenBank mRNA | AK005099.1 | - | 2 | 475 | ||
| AK030173.1 | - | 2 | 481 | |||
| AK031089.1 | - | 2 | 459 | |||
| AK083369.1 | - | 1 | 1479 | |||
| AK087517.1 | - | 2 | 412 | |||
| AK090028.1 | - | 2 | 478 | |||
| AK143635.1 | - | 2 | 482 | |||
| AK173289.1 | - | 2 | 389 | |||
| AK188223.1 | - | 1 | 37 | |||
| BC023852.1 | - | 2 | 465 | 5346910 | ||

Dcaf12
Genomic Map