siRNA Details
| Gene Symbol: Spcs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026911.3 | NP_081187.2 | 4 | 820 | ||
| GenBank mRNA | AK007336.1 | - | 4 | 518 | ||
| AK082928.1 | - | 4 | 519 | |||
| AK088658.1 | - | 2 | 1058 | |||
| BC034270.2 | - | 4 | 695 | |||
| BC049895.1 | - | 4 | 511 | 3991764 | ||

Spcs1
Genomic Map