siRNA Details
| Gene Symbol: Lyzl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026915.2 | NP_081191.1 | 6 | 984 | ||
| GenBank mRNA | BC024932.1 | - | 6 | 984 | 3985345 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lyzl4| Gene Symbol: Lyzl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026915.2 | NP_081191.1 | 6 | 984 | ||
| GenBank mRNA | BC024932.1 | - | 6 | 984 | 3985345 | |