siRNA Details
| Gene Symbol: Zdhhc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026917.5 | NP_081193.2 | 2 | 345 | ||
| GenBank mRNA | AB088044.1 | - | 1 | 111 | ||
| AK007355.1 | - | 2 | 319 | |||
| AK018755.1 | - | 2 | 311 | |||
| AK046166.1 | - | 2 | 345 | |||
| AK080541.1 | - | 2 | 311 | |||
| AK169197.1 | - | 288 | ||||
| AY166672.1 | - | 1 | 111 | |||
| BC005531.1 | - | 2 | 320 | |||
| BC018334.1 | - | 2 | 306 | |||
| BC025126.1 | - | 2 | 330 | 3669723 | ||
| BC052464.1 | - | 2 | 250 | 6834763 | ||
| CJ181332.1 | - | 2 | 345 | |||

Zdhhc3
Genomic Map