siRNA Details
| Gene Symbol: Zdhhc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026917.5 | NP_081193.2 | 7 | 2126 | ||
| GenBank mRNA | AK018755.1 | - | 7 | 2094 | ||
| AK080541.1 | - | 7 | 2093 | |||
| AK187978.1 | - | 1 | 263 | |||
| BC005531.1 | - | 8 | 2153 | |||
| BC025126.1 | - | 7 | 2112 | 3669723 | ||
| BC052464.1 | - | 7 | 2033 | 6834763 | ||

Zdhhc3
Genomic Map