siRNA Details
Gene Symbol: 1810011O10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026931.2 | NP_081207.1 | 1 | 1140 | ||
GenBank mRNA | AK007441.1 | - | 1 | 1141 | ||
BC016562.1 | - | 1 | 1096 | 3154083 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1810011O10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026931.2 | NP_081207.1 | 1 | 1140 | ||
GenBank mRNA | AK007441.1 | - | 1 | 1141 | ||
BC016562.1 | - | 1 | 1096 | 3154083 |