siRNA Details
| Gene Symbol: Sult1c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026935.4 | NP_081211.3 | 8 | 1303 | ||
| GenBank mRNA | AY005469.1 | - | 8 | 1303 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sult1c2| Gene Symbol: Sult1c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026935.4 | NP_081211.3 | 8 | 1303 | ||
| GenBank mRNA | AY005469.1 | - | 8 | 1303 | ||