siRNA Details
| Gene Symbol: Zfp397 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027007.2 | NP_081283.2 | 3 | 716 | ||
| GenBank mRNA | AK013086.1 | - | 3 | 679 | ||
| AK032001.1 | - | 3 | 719 | |||
| AK078454.1 | - | 1 | 2392 | |||
| AY338468.1 | - | 2 | 540 | |||
| BC021456.1 | - | 3 | 700 | |||

Zfp397
Genomic Map