siRNA Details
| Gene Symbol: Hmgb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027036.3 | NP_081312.2 | 1 | 223 | ||
| GenBank mRNA | AK005588.1 | - | 1 | 222 | ||
| BC048744.1 | - | 1 | 221 | |||
| BC061030.1 | - | 1 | 219 | 6775221 | ||
| CA465179.1 | - | 1 | 222 | |||
| CN840046.1 | - | 1 | 226 | |||

Hmgb4
Genomic Map