siRNA Details
| Gene Symbol: Hmgb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027036.3 | NP_081312.2 | 1 | 295 | ||
| GenBank mRNA | AK005588.1 | - | 1 | 294 | ||
| BC048744.1 | - | 1 | 293 | |||
| BC061030.1 | - | 1 | 291 | 6775221 | ||
| BC087934.1 | - | 2 | 251 | |||
| CA465179.1 | - | 1 | 294 | |||
| CN840046.1 | - | 1 | 298 | |||

Hmgb4
Genomic Map