siRNA Details
| Gene Symbol: 1700003M02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027041.4 | NP_081317.4 | 11 | 1469 | ||
| GenBank mRNA | AK005650.1 | - | 3 | 269 | ||
| BC058220.1 | - | 11 | 1441 | 6306249 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700003M02Rik| Gene Symbol: 1700003M02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027041.4 | NP_081317.4 | 11 | 1469 | ||
| GenBank mRNA | AK005650.1 | - | 3 | 269 | ||
| BC058220.1 | - | 11 | 1441 | 6306249 | ||