siRNA Details
| Gene Symbol: Wdfy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111279.1 | NP_001104749.1 | 3 | 311 | ||
| NM_027057.3 | NP_081333.1 | 3 | 311 | |||
| GenBank mRNA | AK005937.1 | - | 3 | 275 | ||
| AK007215.1 | - | 3 | 312 | |||
| AK157204.1 | - | 3 | 270 | |||
| AK159710.1 | - | 3 | 282 | |||
| AK165554.1 | - | 3 | 284 | |||
| AK220391.1 | - | 3 | 245 | |||
| BC025226.1 | - | 3 | 225 | 5011404 | ||

Wdfy1
Genomic Map