siRNA Details
| Gene Symbol: 1700025F22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027074.3 | NP_081350.1 | 4, 5 | 270 | ||
| GenBank mRNA | AK006335.1 | - | 4, 5 | 271 | ||
| BC048529.1 | - | 3, 4 | 198 | 6774736 | ||
| BC061241.1 | - | 4, 5 | 278 | 6743876 | ||

1700025F22Rik
Genomic Map