siRNA Details
| Gene Symbol: DYNLT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291602.1 | NP_001278531.1 | 4 | 507 | ||
| NM_001291603.1 | NP_001278532.1 | 5 | 529 | |||
| NM_006519.3 | NP_006510.1 | 5 | 549 | |||
| GenBank mRNA | AK026669.1 | - | 2 | 2673 | ||
| BC029412.1 | - | 5 | 502 | 4074224 | ||
| BC105588.1 | - | 5 | 498 | 6723966 | ||
| D50663.1 | - | 5 | 483 | |||
| HY017024.1 | - | 4 | 460 | |||
| U56255.1 | - | 5 | 471 | |||

DYNLT1
Genomic Map