siRNA Details
Gene Symbol: Krtap4-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027087.3 | NP_081363.1 | 1 | 748 | ||
GenBank mRNA | AK009035.1 | - | 1 | 749 |
Gene Symbol: Gm11569 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099312.1 | NP_001092782.1 | 1 | 789 |
