siRNA Details
| Gene Symbol: Eqtn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290623.1 | NP_001277552.1 | 4 | 460 | ||
| NM_027089.4 | NP_081365.2 | 4 | 460 | |||
| GenBank mRNA | AB438105.1 | - | 4 | 369 | ||
| AB438106.1 | - | 4 | 369 | |||
| AK006446.1 | - | 4 | 411 | |||
| AK016738.1 | - | 4 | 431 | |||
| BC089517.1 | - | 4 | 427 | 6740450 | ||
| BY716419.1 | - | 4 | 427 | |||
| DQ336137.1 | - | 4 | 411 | |||
| EU828781.1 | - | 4 | 445 | |||
| EU828782.1 | - | 4 | 445 | |||

Eqtn
Genomic Map