siRNA Details
Gene Symbol: Klhdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027117.3 | NP_081393.2 | 10, 11 | 1274 | ||
GenBank mRNA | AK009481.1 | - | 10, 11 | 1189 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Klhdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027117.3 | NP_081393.2 | 10, 11 | 1274 | ||
GenBank mRNA | AK009481.1 | - | 10, 11 | 1189 |