siRNA Details
Gene Symbol: Mfsd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027122.3 | NP_081398.2 | 5 | 1839 | ||
GenBank mRNA | AK089539.1 | - | 5 | 1840 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mfsd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027122.3 | NP_081398.2 | 5 | 1839 | ||
GenBank mRNA | AK089539.1 | - | 5 | 1840 |