siRNA Details
| Gene Symbol: Mfsd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027122.3 | NP_081398.2 | 5 | 2000 | ||
| GenBank mRNA | AK089539.1 | - | 5 | 2001 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mfsd3| Gene Symbol: Mfsd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027122.3 | NP_081398.2 | 5 | 2000 | ||
| GenBank mRNA | AK089539.1 | - | 5 | 2001 | ||