siRNA Details
| Gene Symbol: Fastkd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027123.4 | NP_081399.3 | 7 | 2139 | ||
| GenBank mRNA | AK009264.1 | - | 7 | 897 | ||
| AK031142.1 | - | 7 | 2125 | |||
| AK032530.1 | - | 7 | 2222 | |||
| AK033189.1 | - | 7 | 2140 | |||
| AK036778.1 | - | 7 | 2222 | |||
| AK148902.1 | - | 7 | 2130 | |||
| BC060688.1 | - | 6 | 4852 | 6409274 | ||

Fastkd3
Genomic Map