siRNA Details
| Gene Symbol: Tmbim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027154.5 | NP_081430.3 | 12 | 1885 | ||
| GenBank mRNA | AK077689.1 | - | 12 | 1710 | ||
| AK144110.1 | - | 12 | 1729 | |||
| AK147297.1 | - | 11 | 10593 | |||
| AK152864.1 | - | 12 | 1728 | |||
| AK166926.1 | - | 11 | 2127 | |||
| AK167286.1 | - | 7 | 1184 | |||
| AK170625.1 | - | 12 | 1695 | |||
| AK171102.1 | - | 12 | 1727 | |||
| AK179608.1 | - | 1 | 294 | |||
| AK220296.1 | - | 10 | 2891 | |||
| BC004752.1 | - | 5 | 1287 | 3584518 | ||
| BC018564.1 | - | 1 | 688 | |||

Tmbim1
Genomic Map