siRNA Details
| Gene Symbol: Ms4a6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027209.3 | NP_081485.2 | 5 | 602 | ||
| GenBank mRNA | AF237909.1 | - | 5 | 602 | ||
| AK007613.1 | - | 533 | ||||
| AK075671.1 | - | 5 | 579 | |||
| AK172345.1 | - | 676 | ||||
| BC027425.1 | - | 5 | 579 | 3984299 | ||

Ms4a6b
Genomic Map