siRNA Details
| Gene Symbol: 1810065E05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027239.2 | NP_081515.2 | 3 | 225 | ||
| GenBank mRNA | AK007956.1 | - | 3 | 261 | ||
| BC026941.1 | - | 3 | 197 | 4024443 | ||
| BY092570.1 | - | 3 | 222 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1810065E05Rik| Gene Symbol: 1810065E05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027239.2 | NP_081515.2 | 3 | 225 | ||
| GenBank mRNA | AK007956.1 | - | 3 | 261 | ||
| BC026941.1 | - | 3 | 197 | 4024443 | ||
| BY092570.1 | - | 3 | 222 | |||