siRNA Details
Gene Symbol: FSBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256141.1 | NP_001243070.1 | 1, 2 | 451 | ||
GenBank mRNA | AF007866.1 | - | 1, 2 | 403 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FSBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256141.1 | NP_001243070.1 | 1, 2 | 451 | ||
GenBank mRNA | AF007866.1 | - | 1, 2 | 403 |