siRNA Details
| Gene Symbol: Ptcd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027275.3 | NP_081551.2 | 5 | 303 | ||
| GenBank mRNA | AK032831.1 | - | 5 | 287 | ||
| AK146005.1 | - | 5 | 292 | |||
| AK146172.1 | - | 5 | 304 | |||
| AK172480.1 | - | 5 | 291 | |||
| BC115434.2 | - | 5 | 299 | 40094203 | ||

Ptcd3
Genomic Map