siRNA Details
| Gene Symbol: Ptcd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027275.3 | NP_081551.2 | 18, 19 | 1470 | ||
| GenBank mRNA | AK013131.1 | - | 4, 5 | 249 | ||
| AK146005.1 | - | 18, 19 | 1459 | |||
| AK146172.1 | - | 18, 19 | 1471 | |||
| AK172480.1 | - | 19, 20 | 1535 | |||
| BC026759.1 | - | 2, 3 | 125 | |||
| BC055844.1 | - | 4, 5 | 297 | |||
| BC060687.1 | - | 9, 10 | 691 | |||
| BC084595.1 | - | 9, 10 | 720 | |||
| BC115434.2 | - | 18, 19 | 1466 | 40094203 | ||

Ptcd3
Genomic Map