siRNA Details
| Gene Symbol: Ube2q1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027315.4 | NP_081591.3 | 1, 2 | 437 | ||
| GenBank mRNA | AK009324.1 | - | 19 | |||
| AK146707.1 | - | 1, 2 | 250 | |||
| AY112699.1 | - | 1, 2 | 430 | |||
| AY945937.1 | - | 1, 2 | 334 | |||

Ube2q1
Genomic Map