siRNA Details
| Gene Symbol: Fam162a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027342.1 | NP_081618.1 | 2 | 127 | ||
| GenBank mRNA | AK009946.1 | - | 2 | 127 | ||
| BC010826.1 | - | 2 | 136 | 4219531 | ||
| M97159.1 | - | 98 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam162a| Gene Symbol: Fam162a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027342.1 | NP_081618.1 | 2 | 127 | ||
| GenBank mRNA | AK009946.1 | - | 2 | 127 | ||
| BC010826.1 | - | 2 | 136 | 4219531 | ||
| M97159.1 | - | 98 | ||||