siRNA Details
| Gene Symbol: Afap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027373.2 | NP_081649.1 | 6 | 877 | ||
| GenBank mRNA | AK011153.1 | - | 6 | 705 | ||
| AK080756.1 | - | 6 | 812 | |||
| AK129483.1 | - | 6 | 804 | |||
| AK147706.1 | - | 6 | 769 | |||
| AK148000.1 | - | 6 | 732 | |||
| AK161385.1 | - | 6 | 828 | |||
| BC050814.1 | - | 6 | 877 | 6773872 | ||

Afap1
Genomic Map