siRNA Details
| Gene Symbol: Afap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027373.2 | NP_081649.1 | 6 | 904 | ||
| GenBank mRNA | AK011153.1 | - | 6 | 732 | ||
| AK080756.1 | - | 6 | 839 | |||
| AK129483.1 | - | 6 | 831 | |||
| AK147706.1 | - | 6 | 796 | |||
| AK148000.1 | - | 6 | 759 | |||
| AK161385.1 | - | 6 | 855 | |||
| BC050814.1 | - | 6 | 904 | 6773872 | ||

Afap1
Genomic Map