siRNA Details
| Gene Symbol: Gpatch2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027405.2 | NP_081681.2 | 10 | 3554 | ||
| GenBank mRNA | AK031712.1 | - | 9 | 3460 | ||
| AK129299.1 | - | 10 | 3531 | |||
| AK147916.1 | - | 10 | 3540 | |||
| BC044824.1 | - | 8 | 2779 | |||
| BC048169.1 | - | 10 | 3562 | 6821902 | ||
| BC050782.1 | - | 11 | 3504 | |||
| BC058256.1 | - | 10 | 3554 | 6466603 | ||

Gpatch2l
Genomic Map