siRNA Details
| Gene Symbol: Ints2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027421.2 | NP_081697.2 | 2 | 642 | ||
| GenBank mRNA | AK037055.1 | - | 2 | 615 | ||
| AK129323.1 | - | 2 | 624 | |||
| BC050081.1 | - | 2 | 418 | 30023969 | ||
| BC059056.1 | - | 1 | 494 | 6821329 | ||
| BC118007.1 | - | 2 | 357 | 40086155 | ||
| CF166557.1 | - | 2 | 335 | |||

Ints2
Genomic Map