siRNA Details
| Gene Symbol: Rufy2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027425.3 | NP_081701.2 | 2, 3 | 282 | ||
| GenBank mRNA | AK011847.1 | - | 2, 3 | 255 | ||
| AK045485.1 | - | 2, 3 | 283 | |||
| AK083205.1 | - | 2, 3 | 267 | |||
| AK173195.1 | - | 2, 3 | 243 | |||
| BB643897.1 | - | 2, 3 | 283 | |||
| BC017645.1 | - | 2, 3 | 244 | |||
| BC038265.1 | - | 2, 3 | 223 | |||
| BC137736.1 | - | 2, 3 | 231 | 8860755 | ||
| BC137737.1 | - | 2, 3 | 231 | 8860756 | ||

Rufy2
Genomic Map