siRNA Details
| Gene Symbol: Wdr77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027432.3 | NP_081708.1 | 8 | 862 | ||
| GenBank mRNA | AK012014.1 | - | 8 | 853 | ||
| AK032897.1 | - | 6 | 401 | |||
| AK048488.1 | - | 8 | 885 | |||
| AK050391.1 | - | 8 | 862 | |||
| AK078856.1 | - | 9 | 821 | |||
| AK131608.1 | - | 7 | 773 | |||
| AK161812.1 | - | 8 | 837 | |||
| AK163839.1 | - | 857 | ||||
| AK168006.1 | - | 8 | 864 | |||
| AK168752.1 | - | 8 | 884 | |||
| AK169128.1 | - | 8 | 863 | |||
| BC005755.1 | - | 8 | 817 | 2654151 | ||

Wdr77
Genomic Map