siRNA Details
| Gene Symbol: Idh3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029573.2 | NP_083849.1 | 7 | 698 | ||
| GenBank mRNA | AK003393.1 | - | 4 | 591 | ||
| AK010065.1 | - | 7 | 722 | |||
| AK032787.1 | - | 7 | 722 | |||
| AK047951.1 | - | 7 | 720 | |||
| AK117103.1 | - | 7 | 734 | |||
| AK146717.1 | - | 4 | 562 | |||
| AK150618.1 | - | 7 | 729 | |||
| AK151304.1 | - | 7 | 723 | |||
| AK152353.1 | - | 7 | 732 | |||
| AK153459.1 | - | 7 | 723 | |||
| AK158646.1 | - | 7 | 876 | |||
| AK159051.1 | - | 7 | 795 | |||
| AK159666.1 | - | 7 | 729 | |||
| AK160054.1 | - | 7 | 729 | |||
| AK164038.1 | - | 8 | 825 | |||
| AK167822.1 | - | 7 | 728 | |||
| AK168034.1 | - | 7 | 720 | |||
| AK168049.1 | - | 7 | 723 | |||
| AK168149.1 | - | 7 | 729 | |||
| AK169152.1 | - | 7 | 696 | |||
| BC034273.1 | - | 7 | 688 | 4166193 | ||
| BC049956.1 | - | 7 | 698 | 4212315 | ||

Idh3a
Genomic Map