siRNA Details
| Gene Symbol: Tulp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001103181.1 | NP_001096651.1 | 13 | 3924 | ||
| NM_054040.3 | NP_473381.1 | 13 | 4766 | |||
| GenBank mRNA | AF219945.1 | - | 13 | 4170 | ||
| AK039439.1 | - | 1 | 648 | |||
| AK139719.1 | - | 1055 | ||||
| AK147643.1 | - | 13 | 3864 | |||
| BC008557.1 | - | 1 | 137 | |||
| BC138024.1 | - | 13 | 4101 | 8861043 | ||
| BC138025.1 | - | 13 | 4101 | 8861044 | ||

Tulp4
Genomic Map